Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hjurp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191813.1 | NP_001178742.1 | 4 - 5 | 252 | 80 | |
XM_006245393.2 | XP_006245455.1 | 4 - 5 | 418 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |