Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107615.1 | NP_001101085.1 | 2 - 3 | 1627 | 60 | |
XM_006256129.2 | XP_006256191.1 | 3 - 4 | 2067 | 60 | ||
XM_006256130.2 | XP_006256192.1 | 3 - 4 | 2067 | 60 | ||
XM_006256131.2 | XP_006256193.1 | 3 - 4 | 1776 | 60 | ||
XM_006256132.2 | XP_006256194.1 | 3 - 4 | 1984 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |