Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam92b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772654.1 | XP_008770876.1 | 5 - 6 | 667 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |