Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tysnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108932.1 | NP_001102402.1 | 3 - 4 | 1547 | 90 | |
GenBank mRNA | BC160877.1 | - | 3 - 4 | 1510 | 7373508 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |