Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsb66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181694.2 | NP_859045.2 | 1 - 2 | 48 | 72 | |
XM_006233763.2 | XP_006233825.1 | 1 - 2 | 427 | 72 | ||
GenBank mRNA | BC097268.1 | - | 1 - 2 | 48 | 7457912 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |