Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106566.1 | NP_001100036.1 | 1 - 2 | 48 | 54 | |
XM_006233781.1 | XP_006233843.1 | 1 - 2 | 97 | 54 | ||
GenBank mRNA | FQ231217.1 | - | 1 - 2 | 52 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |