Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024762.1 | NP_001019933.1 | 8 - 9 | 878 | 90 | |
XM_006244686.2 | XP_006244748.1 | 6 - 7 | 707 | 90 | ||
XM_006244689.2 | XP_006244751.1 | 6 - 7 | 658 | 90 | ||
XM_006244690.2 | XP_006244752.1 | 6 - 7 | 724 | 90 | ||
XM_006244691.1 | XP_006244753.1 | 6 - 7 | 725 | 90 | ||
GenBank mRNA | AJ011607.1 | - | 5 - 6 | 562 | 90 | |
BC097454.1 | - | 8 - 9 | 878 | 7456950 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |