Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pth2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031089.2 | NP_112351.1 | 4 - 5 | 619 | 74 | |
GenBank mRNA | U55836.1 | - | 4 - 5 | 619 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |