Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138919.2 | NP_620274.1 | 3 - 4 | 637 | 82 | |
XM_006244736.2 | XP_006244798.1 | 3 - 4 | 590 | 82 | ||
XM_006244737.2 | XP_006244799.1 | 3 - 4 | 535 | 82 | ||
GenBank mRNA | AY017209.1 | - | 3 - 4 | 637 | 82 | |
BC061976.1 | - | 3 - 4 | 632 | 5623807 | 82 | |
FQ220352.1 | - | 3 - 4 | 531 | 82 | ||
U96638.1 | - | 2 - 3 | 403 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |