Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007742.1 | NP_001007743.1 | 7 - 8 | 1060 | 101 | |
XM_006233689.1 | XP_006233751.1 | 7 - 8 | 1087 | 101 | ||
GenBank mRNA | BC083603.1 | - | 7 - 8 | 1060 | 7133164 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |