Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107918.1 | NP_001101388.1 | 9 - 10 | 998 | 72 | |
XM_006237938.2 | XP_006238000.1 | 9 - 10 | 1026 | 72 | ||
XM_006237939.2 | XP_006238001.1 | 9 - 10 | 1028 | 72 | ||
XM_006237940.2 | XP_006238002.1 | 9 - 10 | 1023 | 72 | ||
XM_006237941.2 | XP_006238003.1 | 9 - 10 | 1026 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |