Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107921.1 | NP_001101391.1 | 4 - 5 | 484 | 65 | |
XM_006237973.2 | XP_006238035.1 | 5 - 6 | 384 | 65 | ||
XM_006237974.2 | XP_006238036.1 | 5 - 6 | 560 | 65 | ||
XM_006237975.2 | XP_006238037.1 | 4 - 5 | 314 | 65 | ||
XM_008763620.1 | XP_008761842.1 | 4 - 5 | 1478 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |