Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 5 - 6 | 962 | 67 | |
XM_006251211.2 | XP_006251273.1 | 5 - 6 | 1165 | 67 | ||
XM_006251212.2 | XP_006251274.1 | 5 - 6 | 1165 | 67 | ||
XM_006251213.2 | XP_006251275.1 | 5 - 6 | 1331 | 67 | ||
XM_006251214.2 | XP_006251276.1 | 5 - 6 | 573 | 67 | ||
GenBank mRNA | BC081868.1 | - | 5 - 6 | 962 | 7096894 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |