Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053649.1 | NP_446101.1 | 2 - 3 | 269 | 61 | |
GenBank mRNA | AB065090.1 | - | 2 - 3 | 269 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |