Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077530.1 | NP_001070998.1 | 5 - 6 | 736 | 111 | |
GenBank mRNA | BC105629.1 | - | 5 - 6 | 736 | 7322855 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |