Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Potea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770390.1 | XP_008768612.1 | 11 - 12 | 1412 | 78 | |
XM_008770391.1 | XP_008768613.1 | 11 - 12 | 1412 | 78 | ||
XM_008770392.1 | XP_008768614.1 | 10 - 11 | 1238 | 78 | ||
XM_008770393.1 | XP_008768615.1 | 6 - 7 | 2154 | 78 | ||
GenBank mRNA | AB095021.1 | - | 11 - 12 | 1281 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |