Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044255.1 | NP_001037720.1 | 1 - 2 | 254 | 139 | |
XM_006250675.2 | XP_006250737.1 | 1 - 2 | 286 | 139 | ||
GenBank mRNA | BC099827.1 | - | 1 - 2 | 254 | 7934175 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |