Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025713.1 | NP_001020884.1 | 2 - 3 | 123 | 78 | |
XM_008773568.1 | XP_008771790.1 | 2 - 3 | 104 | 78 | ||
GenBank mRNA | BC097464.1 | - | 2 - 3 | 123 | 7457620 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |