Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109576.1 | NP_001103046.1 | 6 - 7 | 579 | 108 | |
XM_006247737.2 | XP_006247799.1 | 6 - 7 | 1502 | 108 | ||
GenBank mRNA | BC166577.1 | - | 7 - 8 | 604 | 189 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |