Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081980.1 | NP_001075449.1 | 3 - 4 | 299 | 118 | |
XM_006230840.2 | XP_006230902.1 | 3 - 4 | 352 | 118 | ||
GenBank mRNA | BC160813.1 | - | 3 - 4 | 310 | 7385853 | 118 |
FQ226277.1 | - | 3 - 4 | 310 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |