Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024326.1 | NP_001019497.1 | 2 - 3 | 323 | 59 | |
XM_008762820.1 | XP_008761042.1 | 2 - 3 | 751 | 59 | ||
GenBank mRNA | BC088443.1 | - | 2 - 3 | 323 | 7126386 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |