Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024791.1 | NP_001019962.1 | 7 - 8 | 1418 | 110 | |
XM_006247201.2 | XP_006247263.1 | 8 - 9 | 2602 | 110 | ||
XM_006247202.2 | XP_006247264.1 | 8 - 9 | 2567 | 110 | ||
XM_006247203.2 | XP_006247265.1 | 7 - 8 | 2521 | 110 | ||
GenBank mRNA | BC097500.1 | - | 7 - 8 | 1418 | 7462750 | 110 |
Gene Symbol: LOC102555637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247212.1 | XP_006247274.1 | 2 - 3 | 129 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |