Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175765.2 | NP_786941.1 | 8 - 9 | 947 | 108 | |
XM_006238354.2 | XP_006238416.1 | 9 - 10 | 1170 | 108 | ||
XM_008763779.1 | XP_008762001.1 | 9 - 10 | 1170 | 108 | ||
GenBank mRNA | AB285525.1 | - | 850 | 108 | ||
AF339084.1 | - | 8 - 9 | 947 | 108 | ||
BC093606.1 | - | 9 - 10 | 1105 | 7317952 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |