Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106442.1 | NP_001099912.1 | 1 - 2 | 73 | 80 | |
GenBank mRNA | BC168754.1 | - | 1 - 2 | 45 | 6886830 | 80 |
FQ229540.1 | - | 1 - 2 | 45 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |