Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768450.1 | XP_008766672.1 | 23 - 24 | 3768 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |