Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017009.2 | NP_058705.2 | 2 - 3 | 516 | 68 | |
XM_008768261.1 | XP_008766483.1 | 2 - 3 | 582 | 68 | ||
GenBank mRNA | BC088851.1 | - | 2 - 3 | 516 | 7315338 | 68 |
EF094476.1 | - | 2 - 3 | 587 | 68 | ||
L27219.1 | - | 2 - 3 | 520 | 68 | ||
U03700.1 | - | 2 - 3 | 540 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |