Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012602.1 | NP_036734.1 | 6 - 7 | 1954 | 81 | |
XM_006232590.2 | XP_006232652.1 | 7 - 8 | 2219 | 81 | ||
XM_008761128.1 | XP_008759350.1 | 7 - 8 | 2220 | 81 | ||
GenBank mRNA | AF007554.1 | - | 1 - 2 | 156 | 81 | |
BC166505.1 | - | 6 - 7 | 1954 | 7122817 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |