Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107039.1 | NP_001100509.1 | 6 - 7 | 819 | 62 | |
XM_006247191.2 | XP_006247253.1 | 5 - 6 | 643 | 62 | ||
XM_006247192.1 | XP_006247254.1 | 5 - 6 | 547 | 62 | ||
GenBank mRNA | FQ212993.1 | - | 4 - 5 | 450 | 62 | |
FQ215532.1 | - | 5 - 6 | 605 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |