Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133383.1 | NP_596874.1 | 4 - 5 | 486 | 81 | |
GenBank mRNA | AF330051.1 | - | 4 - 5 | 486 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |