Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252478.2 | XP_006252540.1 | 21 - 22 | 2676 | 121 | |
XM_224534.8 | XP_224534.7 | 22 - 23 | 2922 | 121 | ||
GenBank mRNA | FQ214398.1 | - | 1 - 2 | 73 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |