Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008280.1 | NP_001008281.1 | 3 - 4 | 532 | 55 | |
GenBank mRNA | BC086530.1 | - | 3 - 4 | 532 | 7304726 | 55 |
D13623.1 | - | 3 - 4 | 569 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |