Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139115.2 | NP_620815.2 | 1 - 2 | 222 | 69 | |
XM_006246885.2 | XP_006246947.1 | 2 - 3 | 1299 | 69 | ||
XM_006246886.2 | XP_006246948.1 | 2 - 3 | 1301 | 69 | ||
XM_006246887.2 | XP_006246949.1 | 2 - 3 | 1299 | 69 | ||
XM_006246888.2 | XP_006246950.1 | 2 - 3 | 1301 | 69 | ||
XM_006246889.2 | XP_006246951.1 | 2 - 3 | 1299 | 69 | ||
XM_006246890.2 | XP_006246952.1 | 2 - 3 | 1299 | 69 | ||
XM_006246891.2 | XP_006246953.1 | 2 - 3 | 1300 | 69 | ||
GenBank mRNA | BC080241.1 | - | 2 - 3 | 374 | 69 | |
BC098656.1 | - | 1 - 2 | 222 | 7192854 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |