Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105772.1 | NP_001099242.1 | 4 - 5 | 779 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |