Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013975.2 | NP_001013997.2 | 8 - 9 | 992 | 83 | |
XM_006247887.2 | XP_006247949.1 | 8 - 9 | 1225 | 83 | ||
XM_006247888.2 | XP_006247950.1 | 7 - 8 | 1562 | 83 | ||
GenBank mRNA | BC090016.1 | - | 1 - 2 | 374 | 83 | |
CO562343.1 | - | 3 - 4 | 325 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |