Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106710.1 | NP_001100180.1 | 5 - 6 | 3035 | 141 | |
XM_006239793.2 | XP_006239855.1 | 5 - 6 | 3046 | 141 | ||
XM_008764511.1 | XP_008762733.1 | 5 - 6 | 3778 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |