Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185025.1 | NP_001171954.1 | 1 - 2 | 115 | 149 | |
GenBank mRNA | BP486352.1 | - | 1 - 2 | 121 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |