Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012975.1 | NP_037107.1 | 6 - 7 | 560 | 70 | |
XM_008759113.1 | XP_008757335.1 | 6 - 7 | 727 | 70 | ||
GenBank mRNA | M73553.1 | - | 6 - 7 | 560 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |