Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031238.2 | NP_112517.1 | 2 - 3 | 457 | 105 | |
XM_006229480.2 | XP_006229542.1 | 2 - 3 | 245 | 105 | ||
XM_006229481.2 | XP_006229543.1 | 2 - 3 | 338 | 105 | ||
XM_006229483.2 | XP_006229545.1 | 2 - 3 | 444 | 105 | ||
GenBank mRNA | AF227439.1 | - | 2 - 3 | 460 | 105 | |
BC078800.1 | - | 2 - 3 | 460 | 7135194 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |