Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008511.2 | NP_001008511.2 | 6 - 7 | 1126 | 70 | |
XM_006230755.1 | XP_006230817.1 | 6 - 7 | 1239 | 70 | ||
GenBank mRNA | AY280663.1 | - | 5 - 6 | 1020 | 70 | |
BC104701.1 | - | 6 - 7 | 1126 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Rat Estrogen Panel 1700 Inventoried Probe spans exons |