Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039031.1 | NP_001034120.1 | 15 - 16 | 1735 | 115 | |
XM_006231056.2 | XP_006231118.1 | 15 - 16 | 1621 | 115 | ||
XM_006231057.2 | XP_006231119.2 | 15 - 16 | 1904 | 115 | ||
GenBank mRNA | BC098925.1 | - | 15 - 16 | 1735 | 7112371 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |