Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039031.1 | NP_001034120.1 | 2 - 3 | 279 | 70 | |
XM_006231056.2 | XP_006231118.1 | 2 - 3 | 165 | 70 | ||
XM_006231057.2 | XP_006231119.2 | 2 - 3 | 448 | 70 | ||
GenBank mRNA | BC098925.1 | - | 2 - 3 | 279 | 7112371 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |