Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053669.1 | NP_446121.1 | 3 - 4 | 1080 | 86 | |
XM_006249092.2 | XP_006249154.1 | 3 - 4 | 1832 | 86 | ||
XM_006249093.2 | XP_006249155.1 | 3 - 4 | 1833 | 86 | ||
XM_008769112.1 | XP_008767334.1 | 4 - 5 | 1631 | 86 | ||
XM_008769113.1 | XP_008767335.1 | 3 - 4 | 1095 | 86 | ||
GenBank mRNA | AF095576.1 | - | 3 - 4 | 1080 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |