Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105933.1 | NP_001099403.1 | 6 - 7 | 591 | 104 | |
GenBank mRNA | BC166554.1 | - | 6 - 7 | 550 | 7311069 | 104 |
FQ219862.1 | - | 6 - 7 | 565 | 104 | ||
FQ221109.1 | - | 6 - 7 | 104 | |||
FQ232695.1 | - | 6 - 7 | 543 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |