Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107755.1 | NP_001101225.1 | 1 - 2 | 124 | 62 | |
XM_008762061.1 | XP_008760283.1 | 1 - 2 | 241 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus Probe spans exons |