Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234788.2 | XP_006234850.1 | 9 - 10 | 742 | 107 | |
XM_006234789.2 | XP_006234851.1 | 9 - 10 | 722 | 107 | ||
XM_006234790.2 | XP_006234852.1 | 9 - 10 | 767 | 107 | ||
XM_006234791.2 | XP_006234853.1 | 8 - 9 | 624 | 107 | ||
XM_008762112.1 | XP_008760334.1 | 9 - 10 | 799 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |