Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234874.2 | XP_006234936.1 | 9 - 10 | 1645 | 72 | |
XM_006234875.2 | XP_006234937.1 | 8 - 9 | 1552 | 72 | ||
XM_006234876.2 | XP_006234938.1 | 8 - 9 | 1090 | 72 | ||
XM_006234877.1 | XP_006234939.1 | 8 - 9 | 1083 | 72 | ||
XM_008762207.1 | XP_008760429.1 | 5 - 6 | 762 | 72 | ||
XM_008762208.1 | XP_008760430.1 | 6 - 7 | 684 | 72 | ||
XM_008762209.1 | XP_008760431.1 | 5 - 6 | 511 | 72 | ||
XM_008762210.1 | XP_008760432.1 | 5 - 6 | 449 | 72 | ||
XM_230512.7 | XP_230512.4 | 8 - 9 | 1121 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |