Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109606.1 | NP_001103076.1 | 1 - 2 | 119 | 84 | |
XM_006234656.2 | XP_006234718.1 | 1 - 2 | 144 | 84 | ||
XM_006234657.2 | XP_006234719.1 | 1 - 2 | 144 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |