Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014142.1 | NP_001014164.1 | 4 - 5 | 373 | 103 | |
XM_006253159.2 | XP_006253221.1 | 4 - 5 | 363 | 103 | ||
XM_008771265.1 | XP_008769487.1 | 4 - 5 | 524 | 103 | ||
GenBank mRNA | BC083771.1 | - | 4 - 5 | 373 | 7193395 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |