Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005535.2 | NP_001005535.1 | 4 - 5 | 1361 | 135 | |
NM_001190820.1 | NP_001177749.1 | 5 - 6 | 1451 | 135 | ||
GenBank mRNA | BC081871.1 | - | 4 - 5 | 1315 | 7106377 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |