Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024768.1 | NP_001019939.1 | 6 - 7 | 856 | 152 | |
XM_006249881.2 | XP_006249943.1 | 6 - 7 | 951 | 152 | ||
XM_006249883.2 | XP_006249945.1 | 6 - 7 | 882 | 152 | ||
GenBank mRNA | BC097980.1 | - | 6 - 7 | 856 | 7443278 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |